Product Details

SNP ID
rs148937489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32293717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACTTTTCTTTATTTGGGCTCCC[C/T]GTCCTTGTGGTATACTCATCTCACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C6orf10 PubMed Links

Gene Details

Gene
C6orf10
Gene Name
chromosome 6 open reading frame 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286474.1 1162 Missense Mutation CAG,CGG Q317R NP_001273403.1
NM_001286475.1 1162 Missense Mutation CAG,CGG Q303R NP_001273404.1
NM_006781.4 1162 Missense Mutation CAG,CGG Q319R NP_006772.3
XM_017010182.1 1162 Missense Mutation CAG,CGG Q225R XP_016865671.1
XM_017010183.1 1162 Intron XP_016865672.1
XM_017010184.1 1162 Intron XP_016865673.1
XM_017010185.1 1162 Missense Mutation CAG,CGG Q330R XP_016865674.1
Gene
LOC101929163
Gene Name
uncharacterized LOC101929163
There are no transcripts associated with this gene.

View Full Product Details