Product Details

SNP ID
rs149036657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43331564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTCTAAAATACATACCCTCTAT[A/G]TACATTCTAATCTTGTTCATCCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF318 PubMed Links

Gene Details

Gene
ZNF318
Gene Name
zinc finger protein 318
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014345.2 3876 Intron NP_055160.2
XM_011514451.2 3876 Intron XP_011512753.1
XM_011514452.2 3876 UTR 3 XP_011512754.1

View Full Product Details