Product Details

SNP ID
rs149093508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32181432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGTCCCCAGGCCTCCCAGGATC[C/T]CCAGGGCCAGGGCTAGAGTTCCCAG
Phenotype
MIM: 600214 MIM: 603099 MIM: 176311 MIM: 602677
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGER PubMed Links

Gene Details

Gene
AGER
Gene Name
advanced glycosylation end product-specific receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136.4 1185 Missense Mutation GAG,GGG E346G NP_001127.1
NM_001206929.1 1185 Missense Mutation GAG,GGG E362G NP_001193858.1
NM_001206932.1 1185 Missense Mutation GAG,GGG E332G NP_001193861.1
NM_001206934.1 1185 Intron NP_001193863.1
NM_001206936.1 1185 Intron NP_001193865.1
NM_001206940.1 1185 Intron NP_001193869.1
NM_001206954.1 1185 Intron NP_001193883.1
NM_001206966.1 1185 Intron NP_001193895.1
NM_172197.2 1185 Silent Mutation GGA,GGG G294G NP_751947.1
XM_017010328.1 1185 Intron XP_016865817.1
Gene
AGPAT1
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
There are no transcripts associated with this gene.

Gene
GPSM3
Gene Name
G-protein signaling modulator 3
There are no transcripts associated with this gene.

Gene
MIR6833
Gene Name
microRNA 6833
There are no transcripts associated with this gene.

Gene
PBX2
Gene Name
PBX homeobox 2
There are no transcripts associated with this gene.

Gene
RNF5
Gene Name
ring finger protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006913.3 1185 Intron NP_008844.1

View Full Product Details