Product Details

SNP ID
rs149534584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:19838083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGACCTGCAGCTGGCGCTGGAGA[C/T]GCACCCGGCCCTGCTGAGGCAGCCA
Phenotype
MIM: 600581
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ID4 PubMed Links

Gene Details

Gene
ID4
Gene Name
inhibitor of DNA binding 4, HLH protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001546.3 714 Missense Mutation ACG,ATG T110M NP_001537.1

View Full Product Details