Product Details

SNP ID
rs149550435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29427246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGGGCCCCATCAGGAGTGGGTA[A/G]TGGAGTGGGTAGCAAATTGCCAGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR11A1 PubMed Links

Gene Details

Gene
OR11A1
Gene Name
olfactory receptor family 11 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013937.3 487 Silent Mutation CAC,CAT H132H NP_039225.1
Gene
OR5V1
Gene Name
olfactory receptor family 5 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030876.5 487 Intron NP_110503.3
XM_011514929.2 487 Intron XP_011513231.1
XM_011514930.2 487 Intron XP_011513232.1

View Full Product Details