Product Details

SNP ID
rs149954012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:38170128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATGAAGGGCACCTGGCCCCTTC[C/T]GCGAGGTTGGAGCATTATGCCAGGA
Phenotype
MIM: 611237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTBD9 PubMed Links

Gene Details

Gene
BTBD9
Gene Name
BTB domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099272.1 7272 UTR 3 NP_001092742.1
NM_001172418.1 7272 UTR 3 NP_001165889.1
NM_052893.1 7272 UTR 3 NP_443125.1
NM_152733.2 7272 UTR 3 NP_689946.2
XM_011514279.2 7272 UTR 3 XP_011512581.1
XM_011514280.2 7272 Intron XP_011512582.1
XM_011514281.2 7272 Intron XP_011512583.1
XM_011514282.2 7272 Intron XP_011512584.1
XM_017010227.1 7272 Intron XP_016865716.1

View Full Product Details