Product Details

SNP ID
rs150349878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34293496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCTTCCAGCACTTCAGTGACAA[G/T]GAGCACATAGACATACGTCCTGTGC
Phenotype
MIM: 609228
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NUDT3 PubMed Links

Gene Details

Gene
NUDT3
Gene Name
nudix hydrolase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006703.3 746 Missense Mutation ATT,CTT I99L NP_006694.1
Gene
RPS10-NUDT3
Gene Name
RPS10-NUDT3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202470.2 746 Missense Mutation ATT,CTT I218L NP_001189399.1

View Full Product Details