Product Details

SNP ID
rs150491593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167157489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTAAGCCGAGGGTGGACCACAC[A/G]GAGGTACCGGTCCAAAGCCACGGCG
Phenotype
MIM: 602043
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR31 PubMed Links

Gene Details

Gene
GPR31
Gene Name
G protein-coupled receptor 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005299.2 343 Missense Mutation CGT,TGT R115C NP_005290.2
Gene
LOC105378123
Gene Name
uncharacterized LOC105378123
There are no transcripts associated with this gene.

View Full Product Details