Product Details

SNP ID
rs150635890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110392823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCCTGAGGGCATGGACACACTC[A/G]CTCACCAGCCTGGCGGCCTCCAGAG
Phenotype
MIM: 124450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDO PubMed Links

Gene Details

Gene
DDO
Gene Name
D-aspartate oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003649.2 889 Silent Mutation AGC,AGT S354S NP_003640.2
NM_004032.2 889 Silent Mutation AGC,AGT S295S NP_004023.2

View Full Product Details