Product Details

SNP ID
rs150878918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:4996257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGAAGACGGCGCCGAGCCAGTC[C/G]AAGAGCTCCAGAGCCCGGCAGGACA
Phenotype
MIM: 606117
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LYRM4-AS1 PubMed Links

Gene Details

Gene
LYRM4-AS1
Gene Name
LYRM4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RPP40
Gene Name
ribonuclease P/MRP subunit p40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286132.1 658 Silent Mutation TTC,TTG F218L NP_001273061.1
NM_001286133.1 658 Silent Mutation TTC,TTG F199L NP_001273062.1
NM_006638.3 658 Silent Mutation TTC,TTG F241L NP_006629.2
XM_011514253.2 658 Silent Mutation TTC,TTG F241L XP_011512555.1

View Full Product Details