Product Details

SNP ID
rs150925114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32117628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGAAGTGCCGGCAATCAGAGGA[C/G]AGGAAGAGCTGGTCTAGGTCTCTTA
Phenotype
MIM: 600984 MIM: 600985
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATF6B PubMed Links

Gene Details

Gene
ATF6B
Gene Name
activating transcription factor 6 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136153.1 1524 Silent Mutation CTC,CTG L494L NP_001129625.1
NM_004381.4 1524 Silent Mutation CTC,CTG L497L NP_004372.3
Gene
TNXB
Gene Name
tenascin XB
There are no transcripts associated with this gene.

View Full Product Details