Product Details

SNP ID
rs151262083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:90516642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCAGGCGAGATGTATTTTGCTG[A/G]TCCTTTTCATCCTGGTCCAGTTCTG
Phenotype
MIM: 602614
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP3K7 PubMed Links

Gene Details

Gene
MAP3K7
Gene Name
mitogen-activated protein kinase kinase kinase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003188.3 1886 Silent Mutation GAC,GAT D533D NP_003179.1
NM_145331.2 1886 Silent Mutation GAC,GAT D560D NP_663304.1
NM_145332.2 1886 UTR 3 NP_663305.1
NM_145333.2 1886 UTR 3 NP_663306.1
XM_006715553.2 1886 Silent Mutation GAC,GAT D430D XP_006715616.1
XM_017011226.1 1886 Silent Mutation GAC,GAT D403D XP_016866715.1

View Full Product Details