Product Details
- SNP ID
-
rs138007513
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:155462426 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCGAACAAAGAGGACAAGCGGCC[A/G]CGCACGGCCTTTACCGCCGAGCAGC
- Phenotype
-
MIM: 131310
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EN2
PubMed Links
Gene Details
- Gene
- EN2
- Gene Name
- engrailed homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001427.3 |
990 |
Silent Mutation |
CCA,CCG |
P247P |
NP_001418.2 |
View Full Product Details