Product Details

SNP ID
rs138726205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122077906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTTAGCCATGGCTGAAAAGCCA[C/T]TGATGTCCCCATAAGCCACAAGATC
Phenotype
MIM: 605113
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AASS PubMed Links

Gene Details

Gene
AASS
Gene Name
aminoadipate-semialdehyde synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005763.3 2739 Missense Mutation AAT,AGT N865S NP_005754.2
XM_011515725.2 2739 Missense Mutation AAT,AGT N833S XP_011514027.1

View Full Product Details