Product Details
- SNP ID
-
rs138785196
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:5838797 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCCCTGTTCTGCATCTGTGGAAC[C/T]CCTTCTCTGCATTTCTCTTGCTGTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF815P
PubMed Links
Gene Details
- Gene
- ZNF815P
- Gene Name
- zinc finger protein 815, pseudogene
There are no transcripts associated with this gene.
View Full Product Details