Product Details

SNP ID
rs138984388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143444016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTGGCGATAGCATACGGAGCTA[C/G]TGTGAGAAATTCTATCTCTTCCCTC
Phenotype
MIM: 613968
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EPHA1-AS1 PubMed Links

Gene Details

Gene
EPHA1-AS1
Gene Name
EPHA1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TAS2R60
Gene Name
taste 2 receptor member 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177437.1 564 Nonsense Mutation TAC,TAG Y188* NP_803186.1

View Full Product Details