Product Details

SNP ID
rs139685066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:23300900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACTTCTGAGGCAAGAAAATGC[A/C]AGAGACATTTGTGTGATCCAGGTTC
Phenotype
MIM: 608259 MIM: 614624
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGF2BP3 PubMed Links

Gene Details

Gene
IGF2BP3
Gene Name
insulin like growth factor 2 mRNA binding protein 3
There are no transcripts associated with this gene.

Gene
MALSU1
Gene Name
mitochondrial assembly of ribosomal large subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138446.1 350 Silent Mutation GCA,GCC A106A NP_612455.1
XM_017011731.1 350 Silent Mutation GCA,GCC A11A XP_016867220.1

View Full Product Details