Product Details

SNP ID
rs139753421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141764551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAGAGTTTCTAGGGTGATGGT[A/G]TGGATGCTGTTGGGTGCACTGCTCT
Phenotype
MIM: 604868
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAS2R3 PubMed Links

Gene Details

Gene
TAS2R3
Gene Name
taste 2 receptor member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016943.2 455 Silent Mutation GTA,GTG V131V NP_058639.1

View Full Product Details