Product Details

SNP ID
rs140314505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27163709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGGTTGAGCAGTCGAGCCACC[G/T]CGTACCTGCGGTCCCTGGTGAGGTA
Phenotype
MIM: 142957 MIM: 142950 MIM: 142956 MIM: 609688
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HOXA-AS3 PubMed Links

Gene Details

Gene
HOXA-AS3
Gene Name
HOXA cluster antisense RNA 3
There are no transcripts associated with this gene.

Gene
HOXA10
Gene Name
homeobox A10
There are no transcripts associated with this gene.

Gene
HOXA10-AS
Gene Name
HOXA10 antisense RNA
There are no transcripts associated with this gene.

Gene
HOXA10-HOXA9
Gene Name
HOXA10-HOXA9 readthrough
There are no transcripts associated with this gene.

Gene
HOXA7
Gene Name
homeobox A7
There are no transcripts associated with this gene.

Gene
HOXA9
Gene Name
homeobox A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152739.3 786 Missense Mutation GAG,GCG E238A NP_689952.1
Gene
MIR196B
Gene Name
microRNA 196b
There are no transcripts associated with this gene.

View Full Product Details