Product Details

SNP ID
rs141002347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150792077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCAGGGGCTGGGAGCTCTGGCC[A/C]CACAAGTTTCGAAGACCTACTCCCA
Phenotype
MIM: 610334 MIM: 610385
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105375566 PubMed Links

Gene Details

Gene
LOC105375566
Gene Name
uncharacterized LOC105375566
There are no transcripts associated with this gene.

Gene
TMEM176A
Gene Name
transmembrane protein 176A
There are no transcripts associated with this gene.

Gene
TMEM176B
Gene Name
transmembrane protein 176B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101311.1 716 Missense Mutation TGG,TGT W233C NP_001094781.1
NM_001101312.1 716 Missense Mutation TGG,TGT W233C NP_001094782.1
NM_001101314.1 716 Missense Mutation TGG,TGT W196C NP_001094784.1
NM_014020.3 716 Missense Mutation TGG,TGT W233C NP_054739.3
XM_006715933.3 716 Missense Mutation TGG,TGT W196C XP_006715996.2

View Full Product Details