Product Details

SNP ID
rs141088317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100093353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGACATCTCCATTAGCCTGATGG[C/T]TTCATTCACATCTTCTTTCTCCACC
Phenotype
MIM: 602296 MIM: 614729 MIM: 600592 MIM: 612983 MIM: 612150 MIM: 612984
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP4M1 PubMed Links

Gene Details

Gene
AP4M1
Gene Name
adaptor related protein complex 4 mu 1 subunit
There are no transcripts associated with this gene.

Gene
COPS6
Gene Name
COP9 signalosome subunit 6
There are no transcripts associated with this gene.

Gene
MCM7
Gene Name
minichromosome maintenance complex component 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278595.1 2543 Missense Mutation ACC,GCC T457A NP_001265524.1
NM_005916.4 2543 Missense Mutation ACC,GCC T633A NP_005907.3
NM_182776.2 2543 Missense Mutation ACC,GCC T457A NP_877577.1
XM_005250348.3 2543 Missense Mutation ACC,GCC T526A XP_005250405.1
XM_017012217.1 2543 Missense Mutation ACC,GCC T526A XP_016867706.1
Gene
MIR106B
Gene Name
microRNA 106b
There are no transcripts associated with this gene.

Gene
MIR25
Gene Name
microRNA 25
There are no transcripts associated with this gene.

Gene
MIR93
Gene Name
microRNA 93
There are no transcripts associated with this gene.

View Full Product Details