Product Details

SNP ID
rs141620829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:23299535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCCCAAACTTTGTCCGCGGCCT[A/G]CACAGCGAGCCTGGGCTGGAGGAGC
Phenotype
MIM: 614624
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MALSU1 PubMed Links

Gene Details

Gene
MALSU1
Gene Name
mitochondrial assembly of ribosomal large subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138446.1 215 Silent Mutation CTA,CTG L61L NP_612455.1
XM_017011731.1 215 Intron XP_016867220.1

View Full Product Details