Product Details

SNP ID
rs142147025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144398264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCAATCTCAAGGGGACCTTCT[C/T]TCCCAGCCTCAACTCACCTATATTC
Phenotype
MIM: 610934
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOBOX PubMed Links

Gene Details

Gene
NOBOX
Gene Name
NOBOX oogenesis homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080413.3 Intron NP_001073882.3
XM_017011742.1 Intron XP_016867231.1

View Full Product Details