Product Details
- SNP ID
-
rs142162337
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:128801398 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGAATGAACTTGAAGACATGGAA[C/T]GCATTCGGGGAGATTATGAGATGGA
- Phenotype
-
MIM: 611902
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC136
PubMed Links
Gene Details
- Gene
- CCDC136
- Gene Name
- coiled-coil domain containing 136
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001201372.1 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
NP_001188301.1 |
NM_022742.4 |
759 |
Missense Mutation |
CGC,TGC |
R187C |
NP_073579.4 |
XM_005250534.3 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_005250591.1 |
XM_005250538.1 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_005250595.1 |
XM_011516483.1 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_011514785.1 |
XM_011516484.1 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_011514786.1 |
XM_011516485.1 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_011514787.1 |
XM_011516486.1 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_011514788.1 |
XM_011516487.2 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_011514789.1 |
XM_011516488.2 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_011514790.1 |
XM_011516489.1 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_011514791.1 |
XM_011516490.2 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_011514792.1 |
XM_017012532.1 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_016868021.1 |
XM_017012533.1 |
759 |
Missense Mutation |
CGC,TGC |
R187C |
XP_016868022.1 |
XM_017012534.1 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_016868023.1 |
XM_017012535.1 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_016868024.1 |
XM_017012536.1 |
759 |
Missense Mutation |
CGC,TGC |
R237C |
XP_016868025.1 |
View Full Product Details