Product Details

SNP ID
rs143147289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:42910103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTCTTTGAAAATAAAGTGGCA[A/G]CCTCCATAGCTGAGGGCAGATACAT
Phenotype
MIM: 176842
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf25 PubMed Links

Gene Details

Gene
C7orf25
Gene Name
chromosome 7 open reading frame 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099858.1 995 Silent Mutation GGC,GGT G324G NP_001093328.1
NM_024054.2 995 Silent Mutation GGC,GGT G266G NP_076959.2
XM_005249844.3 995 Silent Mutation GGC,GGT G266G XP_005249901.1
XM_017012596.1 995 Silent Mutation GGC,GGT G280G XP_016868085.1
XM_017012597.1 995 Silent Mutation GGC,GGT G266G XP_016868086.1
Gene
PSMA2
Gene Name
proteasome subunit alpha 2
There are no transcripts associated with this gene.

View Full Product Details