Product Details

SNP ID
rs143159480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99839158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACAGAGAATGTAATGAAAAATA[C/G]GGAGAAATGTGGGGGTGAGTATTCT
Phenotype
MIM: 606534
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYP3A43 PubMed Links

Gene Details

Gene
CYP3A43
Gene Name
cytochrome P450 family 3 subfamily A member 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278921.1 233 Missense Mutation ACG,AGG T109R NP_001265850.1
NM_022820.4 233 Silent Mutation TAC,TAG Y68* NP_073731.1
NM_057095.2 233 Silent Mutation TAC,TAG Y68* NP_476436.1
NM_057096.3 233 Silent Mutation TAC,TAG Y68* NP_476437.1
XM_017012542.1 233 UTR 5 XP_016868031.1
XM_017012543.1 233 UTR 5 XP_016868032.1
XM_017012544.1 233 Intron XP_016868033.1
XM_017012545.1 233 Intron XP_016868034.1

View Full Product Details