Product Details

SNP ID
rs143351272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100888913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGGTTGTAGAAGGAGTTGGGG[G/T]CAAAGGCTGCACTCACCTCTTGCCA
Phenotype
MIM: 100740 MIM: 614469 MIM: 611481
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACHE PubMed Links

Gene Details

Gene
ACHE
Gene Name
acetylcholinesterase (Cartwright blood group)
There are no transcripts associated with this gene.

Gene
SRRT
Gene Name
serrate, RNA effector molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128852.1 806 Intron NP_001122324.1
NM_001128853.1 806 Intron NP_001122325.1
NM_001128854.1 806 Intron NP_001122326.1
NM_015908.5 806 Intron NP_056992.4
XM_005250405.2 806 Intron XP_005250462.1
XM_005250406.2 806 Intron XP_005250463.1
XM_005250407.2 806 Intron XP_005250464.1
XM_005250408.2 806 Intron XP_005250465.1
XM_017012290.1 806 Intron XP_016867779.1
XM_017012291.1 806 Intron XP_016867780.1
Gene
UFSP1
Gene Name
UFM1 specific peptidase 1 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015072.3 806 Missense Mutation GAC,GCC D120A NP_001015072.2

View Full Product Details