Product Details

SNP ID
rs143413842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143286126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTGTTTGCCTATCTCCTGGTC[C/T]GGTTTCTGGAATGGGGGCTTCGGTC
Phenotype
MIM: 600639 MIM: 616524
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP2 PubMed Links

Gene Details

Gene
CASP2
Gene Name
caspase 2
There are no transcripts associated with this gene.

Gene
LOC105375545
Gene Name
uncharacterized LOC105375545
There are no transcripts associated with this gene.

Gene
TMEM139
Gene Name
transmembrane protein 139
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242773.2 262 Missense Mutation CGG,TGG R57W NP_001229702.1
NM_001242774.2 262 Missense Mutation CGG,TGG R57W NP_001229703.1
NM_001242775.2 262 Missense Mutation CGG,TGG R57W NP_001229704.1
NM_001282876.1 262 Missense Mutation CGG,TGG R57W NP_001269805.1
NM_001282877.1 262 Missense Mutation CGG,TGG R57W NP_001269806.1
NM_153345.3 262 Missense Mutation CGG,TGG R57W NP_699176.1

View Full Product Details