Product Details

SNP ID
rs143569274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100122551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGACTATAGTGTTCACGCTGAG[C/T]GCAAGGGCTCACTGAGATATGCCAA
Phenotype
MIM: 610047 MIM: 602955
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNPY4 PubMed Links

Gene Details

Gene
CNPY4
Gene Name
canopy FGF signaling regulator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152755.1 419 Missense Mutation CGC,TGC R106C NP_689968.1
Gene
LAMTOR4
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 4
There are no transcripts associated with this gene.

Gene
MBLAC1
Gene Name
metallo-beta-lactamase domain containing 1
There are no transcripts associated with this gene.

Gene
TAF6
Gene Name
TATA-box binding protein associated factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190415.1 419 Intron NP_001177344.1
NM_005641.3 419 Intron NP_005632.1
NM_139315.2 419 Intron NP_647476.1

View Full Product Details