Product Details

SNP ID
rs143632843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93425080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGATTACTGTTGCAAATTCACA[A/G]ATTAATTTCAGGTGCCAGTAACGAT
Phenotype
MIM: 114131
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CALCR PubMed Links

Gene Details

Gene
CALCR
Gene Name
calcitonin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164737.1 2984 UTR 3 NP_001158209.1
NM_001164738.1 2984 UTR 3 NP_001158210.1
NM_001742.3 2984 UTR 3 NP_001733.1

View Full Product Details