Product Details
- SNP ID
-
rs143927692
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:24800257 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTCCAAATAGGTGCCGTTGCTCA[A/C]CCAAGAGTCATCGTCGGATTTCCTG
- Phenotype
-
MIM: 606732
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OSBPL3
PubMed Links
Gene Details
- Gene
- OSBPL3
- Gene Name
- oxysterol binding protein like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015550.3 |
2964 |
Missense Mutation |
GTG,TTG |
V864L |
NP_056365.1 |
NM_145320.2 |
2964 |
Missense Mutation |
GTG,TTG |
V833L |
NP_663160.1 |
NM_145321.2 |
2964 |
Missense Mutation |
GTG,TTG |
V828L |
NP_663161.1 |
NM_145322.2 |
2964 |
Missense Mutation |
GTG,TTG |
V797L |
NP_663162.1 |
XM_005249698.3 |
2964 |
Missense Mutation |
GTG,TTG |
V864L |
XP_005249755.1 |
XM_006715681.3 |
2964 |
Missense Mutation |
GTG,TTG |
V833L |
XP_006715744.1 |
XM_006715682.3 |
2964 |
Missense Mutation |
GTG,TTG |
V828L |
XP_006715745.1 |
XM_006715683.3 |
2964 |
Missense Mutation |
GTG,TTG |
V797L |
XP_006715746.1 |
XM_006715684.3 |
2964 |
Missense Mutation |
GTG,TTG |
V511L |
XP_006715747.1 |
XM_011515258.2 |
2964 |
Missense Mutation |
GTG,TTG |
V864L |
XP_011513560.1 |
XM_011515259.2 |
2964 |
Missense Mutation |
GTG,TTG |
V820L |
XP_011513561.1 |
XM_017011948.1 |
2964 |
Missense Mutation |
GTG,TTG |
V820L |
XP_016867437.1 |
XM_017011949.1 |
2964 |
Missense Mutation |
GTG,TTG |
V784L |
XP_016867438.1 |
View Full Product Details