Product Details

SNP ID
rs144045718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27172081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTGAACAGAAACTCCTTCTCCA[G/T]CTCCAGTGTCTGGTGCTTCGTGTAG
Phenotype
MIM: 142957 MIM: 142958 MIM: 142956 MIM: 609688
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HOXA10 PubMed Links

Gene Details

Gene
HOXA10
Gene Name
homeobox A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018951.3 1081 Missense Mutation ATG,CTG M351L NP_061824.3
Gene
HOXA10-AS
Gene Name
HOXA10 antisense RNA
There are no transcripts associated with this gene.

Gene
HOXA10-HOXA9
Gene Name
HOXA10-HOXA9 readthrough
There are no transcripts associated with this gene.

Gene
HOXA11
Gene Name
homeobox A11
There are no transcripts associated with this gene.

Gene
HOXA9
Gene Name
homeobox A9
There are no transcripts associated with this gene.

Gene
MIR196B
Gene Name
microRNA 196b
There are no transcripts associated with this gene.

View Full Product Details