Product Details

SNP ID
rs144187592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143443901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGATGCTCTTCAGCTCTGTAG[A/G]GCTCTCCAGCTTCACCACCATTCTA
Phenotype
MIM: 613968
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPHA1-AS1 PubMed Links

Gene Details

Gene
EPHA1-AS1
Gene Name
EPHA1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TAS2R60
Gene Name
taste 2 receptor member 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177437.1 449 Missense Mutation GAG,GGG E150G NP_803186.1

View Full Product Details