Product Details
- SNP ID
-
rs144308673
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:38317468 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTACGAGCCCCAAGGACTGCCAGCG[G/T]TGATGTGTGGAGCAGTGACAGCATG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TRGV9
PubMed Links
Gene Details
- Gene
- TRGV9
- Gene Name
- T cell receptor gamma variable 9
There are no transcripts associated with this gene.
View Full Product Details