Product Details

SNP ID
rs144364133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:156949921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACGCGACTTCGGAAGGCGAGGCT[C/T]CCGGGGGTTGCGAGGGGCGCGGCGC
Phenotype
MIM: 611269
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOM1 PubMed Links

Gene Details

Gene
NOM1
Gene Name
nucleolar protein with MIF4G domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138400.1 199 Missense Mutation CCC,TCC P62S NP_612409.1

View Full Product Details