Product Details
- SNP ID
-
rs144694231
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:25226655 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGTTTTTGATCGGGATTCTGGAT[G/T]TCTTGGTGCTGGCAGGTCATGGAGT
- Phenotype
-
MIM: 616984
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NPVF
PubMed Links
Gene Details
- Gene
- NPVF
- Gene Name
- neuropeptide VF precursor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022150.3 |
557 |
Missense Mutation |
GAA,GAC |
E170D |
NP_071433.3 |
View Full Product Details