Product Details

SNP ID
rs145643348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100888856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACTTCGTCCTCAGTCCAAGGTG[C/T]GCTGCTGCTGTTGGGAGCTAAGGCT
Phenotype
MIM: 100740 MIM: 614469 MIM: 611481
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACHE PubMed Links

Gene Details

Gene
ACHE
Gene Name
acetylcholinesterase (Cartwright blood group)
There are no transcripts associated with this gene.

Gene
SRRT
Gene Name
serrate, RNA effector molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128852.1 863 Intron NP_001122324.1
NM_001128853.1 863 Intron NP_001122325.1
NM_001128854.1 863 Intron NP_001122326.1
NM_015908.5 863 Intron NP_056992.4
XM_005250405.2 863 Intron XP_005250462.1
XM_005250406.2 863 Intron XP_005250463.1
XM_005250407.2 863 Intron XP_005250464.1
XM_005250408.2 863 Intron XP_005250465.1
XM_017012290.1 863 Intron XP_016867779.1
XM_017012291.1 863 Intron XP_016867780.1
Gene
UFSP1
Gene Name
UFM1 specific peptidase 1 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015072.3 863 Missense Mutation CAC,CGC H139R NP_001015072.2

View Full Product Details