Product Details

SNP ID
rs145999869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:45724485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTCAGATGAAATCAGTTCCTTA[C/T]AGAGAGAAGTTAAAAAAATCATTAT
Phenotype
MIM: 103072 MIM: 611563
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADCY1 PubMed Links

Gene Details

Gene
ADCY1
Gene Name
adenylate cyclase 1
There are no transcripts associated with this gene.

Gene
SEPT7P2
Gene Name
septin 7 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details