Product Details

SNP ID
rs146008713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:56011816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAATCAGTGATATACCATTTGGC[A/G]TTATCCTTGACTTGTTGCCTGATCA
Phenotype
MIM: 172480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSPH PubMed Links

Gene Details

Gene
PSPH
Gene Name
phosphoserine phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004577.3 1353 Silent Mutation AAC,AAT N208N NP_004568.2
XM_005271773.1 1353 Silent Mutation AAC,AAT N208N XP_005271830.1
XM_005271774.1 1353 Silent Mutation AAC,AAT N208N XP_005271831.1
XM_005271775.1 1353 Silent Mutation AAC,AAT N208N XP_005271832.1
XM_005271776.1 1353 Silent Mutation AAC,AAT N208N XP_005271833.1
XM_006715760.1 1353 Silent Mutation AAC,AAT N208N XP_006715823.1
XM_006715761.1 1353 Silent Mutation AAC,AAT N163N XP_006715824.1
XM_011515461.1 1353 Silent Mutation AAC,AAT N208N XP_011513763.1
XM_017012466.1 1353 Silent Mutation AAC,AAT N208N XP_016867955.1
XM_017012467.1 1353 Silent Mutation AAC,AAT N208N XP_016867956.1
XM_017012468.1 1353 Silent Mutation AAC,AAT N163N XP_016867957.1
XM_017012469.1 1353 Silent Mutation AAC,AAT N163N XP_016867958.1

View Full Product Details