Product Details

SNP ID
rs146024949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107180008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATTGGAGTTGCTGCAGTGTAAT[A/G]AGAATTTGCCATCTTCACCTGGATA
Phenotype
MIM: 616714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HBP1 PubMed Links

Gene Details

Gene
HBP1
Gene Name
HMG-box transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244262.1 301 Missense Mutation AAG,GAG K49E NP_001231191.1
NM_012257.3 301 Missense Mutation AAG,GAG K39E NP_036389.2
XM_005250266.2 301 Missense Mutation AAG,GAG K39E XP_005250323.1
XM_005250267.2 301 Missense Mutation AAG,GAG K39E XP_005250324.1
XM_017011967.1 301 Missense Mutation AAG,GAG K39E XP_016867456.1

View Full Product Details