Product Details

SNP ID
rs146143218
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141778562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTGTAGGAATCATTATGAATC[C/T]GTTTATTACAGTGGTCAATTGCAAA
Phenotype
MIM: 604869
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TAS2R4 PubMed Links

Gene Details

Gene
TAS2R4
Gene Name
taste 2 receptor member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016944.1 74 Missense Mutation CCG,CTG P25L NP_058640.1

View Full Product Details