Product Details

SNP ID
rs146219058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140411911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACAGCTGGCCAGGAGCGCTTCC[A/G]CACCATCACCCAAAGCTACTACCGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB19 PubMed Links

Gene Details

Gene
RAB19
Gene Name
RAB19, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008749.2 633 Missense Mutation CAC,CGC H80R NP_001008749.2
XM_005249996.3 633 Missense Mutation CAC,CGC H80R XP_005250053.1
XM_005249997.3 633 Missense Mutation CAC,CGC H68R XP_005250054.1
XM_005249998.3 633 Missense Mutation CAC,CGC H66R XP_005250055.1
XM_017012207.1 633 Missense Mutation CAC,CGC H80R XP_016867696.1
Gene
SLC37A3
Gene Name
solute carrier family 37 member 3
There are no transcripts associated with this gene.

View Full Product Details