Product Details

SNP ID
rs146266307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100889066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCCCAGCAAGGCCTTGGACCTG[C/G]CATCTGCGTCCCCCCCAACCATGAC
Phenotype
MIM: 100740 MIM: 614469 MIM: 611481
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACHE PubMed Links

Gene Details

Gene
ACHE
Gene Name
acetylcholinesterase (Cartwright blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000665.4 653 Intron NP_000656.1
NM_001282449.1 653 Intron NP_001269378.1
NM_001302621.1 653 Intron NP_001289550.1
NM_001302622.1 653 Intron NP_001289551.1
NM_015831.2 653 Intron NP_056646.1
XM_006715995.2 653 Intron XP_006716058.1
XM_011516225.1 653 Intron XP_011514527.1
XM_011516226.1 653 Intron XP_011514528.1
XM_011516228.2 653 Intron XP_011514530.1
XM_011516229.2 653 Intron XP_011514531.1
XM_017012219.1 653 Intron XP_016867708.1
XM_017012220.1 653 Intron XP_016867709.1
Gene
SRRT
Gene Name
serrate, RNA effector molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128852.1 653 Intron NP_001122324.1
NM_001128853.1 653 Intron NP_001122325.1
NM_001128854.1 653 Intron NP_001122326.1
NM_015908.5 653 Intron NP_056992.4
XM_005250405.2 653 Intron XP_005250462.1
XM_005250406.2 653 Intron XP_005250463.1
XM_005250407.2 653 Intron XP_005250464.1
XM_005250408.2 653 Intron XP_005250465.1
XM_017012290.1 653 Intron XP_016867779.1
XM_017012291.1 653 Intron XP_016867780.1
Gene
UFSP1
Gene Name
UFM1 specific peptidase 1 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015072.3 653 Missense Mutation GCC,GGC A69G NP_001015072.2

View Full Product Details