Product Details

SNP ID
rs146301759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127582256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCAGGGCTGCAACCTCCTCAC[A/G]ATGCCGTTCGCCCTCCTCCAGCAGC
Phenotype
MIM: 103188 MIM: 607418
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARF5 PubMed Links

Gene Details

Gene
ARF5
Gene Name
ADP ribosylation factor 5
There are no transcripts associated with this gene.

Gene
GCC1
Gene Name
GRIP and coiled-coil domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024523.5 2504 Missense Mutation NP_078799.2

View Full Product Details