Product Details

SNP ID
hCV173232511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73831244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGTAATGGGCATCGCGCTGGCC[A/G]TCCTGGGCTGGCTGGCCGTCATGCT
Phenotype
MIM: 602909 MIM: 612546
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLDN4 PubMed Links

Gene Details

Gene
CLDN4
Gene Name
claudin 4
There are no transcripts associated with this gene.

Gene
WBSCR27
Gene Name
Williams Beuren syndrome chromosome region 27
There are no transcripts associated with this gene.

View Full Product Details