Product Details

SNP ID
rs146894810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19698119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTATTTTGTATAAAATGGTATT[G/T]TAACAATACTGGCTTTCCCCAAATT
Phenotype
MIM: 608312
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR3146 PubMed Links

Gene Details

Gene
MIR3146
Gene Name
microRNA 3146
There are no transcripts associated with this gene.

Gene
TWISTNB
Gene Name
TWIST neighbor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002926.1 1235 UTR 3 NP_001002926.1

View Full Product Details