Product Details

SNP ID
rs146916223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151048979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCGATGCACGGGCTGGGCCAC[A/G]TCTTCGGGCCAGGCAGCCTGAGCCT
Phenotype
MIM: 605464 MIM: 611741 MIM: 123831 MIM: 109280
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCB8 PubMed Links

Gene Details

Gene
ABCB8
Gene Name
ATP binding cassette subfamily B member 8
There are no transcripts associated with this gene.

Gene
ASIC3
Gene Name
acid sensing ion channel subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004769.3 688 Missense Mutation ATC,GTC I32V NP_004760.1
NM_020321.3 688 Missense Mutation ATC,GTC I32V NP_064717.1
NM_020322.3 688 Missense Mutation ATC,GTC I32V NP_064718.1
Gene
CDK5
Gene Name
cyclin dependent kinase 5
There are no transcripts associated with this gene.

Gene
SLC4A2
Gene Name
solute carrier family 4 member 2
There are no transcripts associated with this gene.

View Full Product Details