Product Details

SNP ID
rs147152815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2360792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTGTAGTGGACAATGTCCCTCA[C/G]GTGGGACCCGACCGACTTGAGAAAC
Phenotype
MIM: 603917 MIM: 614905
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EIF3B PubMed Links

Gene Details

Gene
EIF3B
Gene Name
eukaryotic translation initiation factor 3 subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037283.1 847 Silent Mutation CAC,CAG H194Q NP_001032360.1
NM_003751.3 847 Silent Mutation CAC,CAG H194Q NP_003742.2
XM_011515599.1 847 Missense Mutation CAC,CAG H194Q XP_011513901.1
XM_011515600.1 847 Missense Mutation CAC,CAG H194Q XP_011513902.1
XM_011515601.1 847 Missense Mutation CAC,CAG H155Q XP_011513903.1
XM_017012752.1 847 Missense Mutation CAC,CAG H194Q XP_016868241.1
XM_017012753.1 847 Missense Mutation CAC,CAG H194Q XP_016868242.1
Gene
SNX8
Gene Name
sorting nexin 8
There are no transcripts associated with this gene.

View Full Product Details