Product Details

SNP ID
rs147247886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:55771897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTGATTTTTATTTTTGTAGGCA[C/T]TTGTCTTGGTTATAAGAGGGCAGGG
Phenotype
MIM: 604239
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSPHP1 PubMed Links

Gene Details

Gene
PSPHP1
Gene Name
phosphoserine phosphatase pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details