Product Details
- SNP ID
-
rs147351218
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:56068524 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCATGGTCCAACTGCAGGGTGGGA[C/G]ATTCCTGATGGGAACAAATTCTCCA
- Phenotype
-
MIM: 104613
MIM: 607940
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCT6A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7808729] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCT6A
- Gene Name
- chaperonin containing TCP1 subunit 6A
There are no transcripts associated with this gene.
- Gene
- SNORA15
- Gene Name
- small nucleolar RNA, H/ACA box 15
There are no transcripts associated with this gene.
- Gene
- SUMF2
- Gene Name
- sulfatase modifying factor 2
View Full Product Details